Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003314.1 | NP_001003314.1 | 2 - 3 | 330 | 152 | |
GenBank mRNA | J04067.1 | - | 2 - 3 | 330 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |