Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC11C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003312.1 | NP_001003312.1 | 297 | 93 | ||
GenBank mRNA | J05069.1 | - | 297 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |