Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLIPRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003319.2 | NP_001003319.2 | 6 - 7 | 601 | 123 | |
GenBank mRNA | DN356225.1 | - | 5 - 6 | 588 | 123 | |
DN356885.1 | - | 2 - 3 | 135 | 123 | ||
M35302.1 | - | 6 - 7 | 607 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |