Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003298.1 | NP_001003298.1 | 8 - 9 | 2147 | 96 | |
XM_005621304.2 | XP_005621361.1 | 8 - 9 | 2068 | 96 | ||
XM_005621305.3 | XP_005621362.1 | 8 - 9 | 2010 | 96 | ||
GenBank mRNA | U32086.1 | - | 8 - 9 | 2147 | 96 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Canis familiaris Probe spans exons |