Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003222.1 | NP_001003222.1 | 8 - 9 | 760 | 155 | |
XM_022426495.1 | XP_022282203.1 | 6 - 7 | 1164 | 155 | ||
GenBank mRNA | U83905.1 | - | 8 - 9 | 760 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |