Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003222.1 | NP_001003222.1 | 9 - 10 | 867 | 134 | |
XM_022426495.1 | XP_022282203.1 | 7 - 8 | 1271 | 134 | ||
GenBank mRNA | U83905.1 | - | 9 - 10 | 867 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |