Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002993.2 | NP_001002993.2 | 296 | 75 | ||
XM_022422515.1 | XP_022278223.1 | 296 | 75 | |||
GenBank mRNA | U91844.2 | - | 296 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |