Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003272.1 | NP_001003272.1 | 16 - 17 | 1803 | 72 | |
XM_005623180.3 | XP_005623237.1 | 15 - 16 | 2003 | 72 | ||
XM_005623181.3 | XP_005623238.1 | 15 - 16 | 1609 | 72 | ||
GenBank mRNA | X16318.1 | - | 1803 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |