Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003287.1 | NP_001003287.1 | 2 - 3 | 263 | 71 | |
GenBank mRNA | X53564.1 | - | 208 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |