Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003277.3 | NP_001003277.2 | 2 - 3 | 684 | 141 | |
GenBank mRNA | X56387.1 | - | 323 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |