Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003267.1 | NP_001003267.1 | 9 - 10 | 966 | 131 | |
XM_014109166.2 | XP_013964641.1 | 9 - 10 | 955 | 131 | ||
GenBank mRNA | X63678.1 | - | 966 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |