Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003258.2 | NP_001003258.1 | 3 - 4 | 461 | 75 | |
XM_022417332.1 | XP_022273040.1 | 3 - 4 | 618 | 75 | ||
GenBank mRNA | X76392.1 | - | 462 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |