Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003227.1 | NP_001003227.1 | 6 - 7 | 1609 | 52 | |
XM_005636676.3 | XP_005636733.1 | 8 - 9 | 1874 | 52 | ||
XM_014108245.2 | XP_013963720.1 | 6 - 7 | 1800 | 52 | ||
XM_014108246.2 | XP_013963721.1 | 7 - 8 | 1776 | 52 | ||
XM_022410843.1 | XP_022266551.1 | 7 - 8 | 1802 | 52 | ||
GenBank mRNA | X97226.1 | - | 1609 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |