Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003112.1 | NP_001003112.1 | 3 - 4 | 633 | 134 | |
XM_005638170.3 | XP_005638227.1 | 3 - 4 | 668 | 134 | ||
XM_005638171.3 | XP_005638228.1 | 3 - 4 | 378 | 134 | ||
GenBank mRNA | X99145.2 | - | 633 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |