Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003080.1 | NP_001003080.1 | 1 | 337 | 146 | |
GenBank mRNA | AF285177.1 | - | 1 | 337 | 146 |
Gene Symbol: FTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193656.1 | NP_001180585.1 | 1 | 188 | 146 | |
XM_022405806.1 | XP_022261514.1 | 2 | 211 | 146 | ||
GenBank mRNA | AB175610.1 | - | 1 | 188 | 146 | |
AB175611.1 | - | 1 | 156 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Amplicon spans exons and probe does not span exons |