Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4A38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048017.1 | NP_001041482.1 | 1 - 2 | 248 | 94 | |
XM_005628834.3 | XP_005628891.1 | 1 - 2 | 268 | 94 | ||
XM_005628835.3 | XP_005628892.1 | 1 - 2 | 268 | 94 | ||
GenBank mRNA | DQ138951.1 | - | 1 - 2 | 248 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons |