Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204843.1 | NP_001191772.1 | 3 - 4 | 254 | 70 | |
XM_022423974.1 | XP_022279682.1 | 3 - 4 | 416 | 70 | ||
GenBank mRNA | BM537539.1 | - | 3 - 4 | 258 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |