Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630308.3 | XP_005630365.1 | 4 - 5 | 551 | 68 | |
XM_022404611.1 | XP_022260319.1 | 3 - 4 | 503 | 68 | ||
XM_532921.4 | XP_532921.1 | 2 - 3 | 301 | 68 | ||
XM_857398.5 | XP_862491.1 | 2 - 3 | 1436 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |