Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003638821.2 | XP_003638869.1 | 2 - 3 | 355 | 105 | |
XM_005616315.1 | XP_005616372.1 | 2 - 3 | 337 | 105 | ||
XM_859219.4 | XP_864312.1 | 2 - 3 | 360 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |