Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048101.2 | NP_001041566.1 | 2 - 3 | 395 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |