Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114701.2 | XP_013970176.1 | 6 - 7 | 812 | 113 | |
XM_022420269.1 | XP_022275977.1 | 6 - 7 | 841 | 113 | ||
XM_022420270.1 | XP_022275978.1 | 6 - 7 | 788 | 113 | ||
XM_022420271.1 | XP_022275979.1 | 6 - 7 | 920 | 113 | ||
XM_022420272.1 | XP_022275980.1 | 6 - 7 | 915 | 113 | ||
XM_847090.5 | XP_852183.1 | 6 - 7 | 732 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |