Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022405520.1 | XP_022261228.1 | 5 - 6 | 242 | 67 | |
XM_845975.5 | XP_851068.1 | 5 - 6 | 349 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |