Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048095.1 | NP_001041560.1 | 2 - 3 | 248 | 144 | |
XM_005618876.2 | XP_005618933.1 | 3 - 4 | 368 | 144 | ||
GenBank mRNA | DQ143043.1 | - | 2 - 3 | 248 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |