Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFE2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624612.3 | XP_005624669.1 | 1450 | 64 | ||
XM_014116620.2 | XP_013972095.1 | 1337 | 64 | |||
XM_022423058.1 | XP_022278766.1 | 1336 | 64 | |||
XM_846772.5 | XP_851865.1 | 1453 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris |