Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAXX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627201.3 | XP_005627258.1 | 7 - 8 | 2336 | 78 | |
XM_005627204.3 | XP_005627261.1 | 7 - 8 | 2328 | 78 | ||
XM_005627205.3 | XP_005627262.1 | 6 - 7 | 1961 | 78 | ||
XM_022425929.1 | XP_022281637.1 | 6 - 7 | 2145 | 78 | ||
XM_022425930.1 | XP_022281638.1 | 7 - 8 | 2334 | 78 | ||
XM_022425931.1 | XP_022281639.1 | 7 - 8 | 2332 | 78 | ||
XM_022425932.1 | XP_022281640.1 | 7 - 8 | 2275 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |