Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EWSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290126.1 | NP_001277055.1 | 4 - 5 | 288 | 104 | |
XM_003433395.4 | XP_003433443.1 | 4 - 5 | 354 | 104 | ||
XM_022410155.1 | XP_022265863.1 | 4 - 5 | 357 | 104 | ||
XM_022410156.1 | XP_022265864.1 | 4 - 5 | 357 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |