Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271792.1 | NP_001258721.1 | 1 - 2 | 140 | 64 | |
NM_001271793.1 | NP_001258722.1 | 1 - 2 | 140 | 64 | ||
NM_001271794.1 | NP_001258723.1 | 1 - 2 | 140 | 64 | ||
NM_001271795.1 | NP_001258724.1 | 1 - 2 | 140 | 64 | ||
XM_005633292.2 | XP_005633349.1 | 2 - 3 | 312 | 64 | ||
XM_005633293.2 | XP_005633350.1 | 3 - 4 | 331 | 64 | ||
XM_005633294.2 | XP_005633351.1 | 2 - 3 | 313 | 64 | ||
XM_005633295.2 | XP_005633352.1 | 2 - 3 | 314 | 64 | ||
XM_022407426.1 | XP_022263134.1 | 2 - 3 | 314 | 64 | ||
XM_022407427.1 | XP_022263135.1 | 3 - 4 | 333 | 64 | ||
XM_022407428.1 | XP_022263136.1 | 2 - 3 | 314 | 64 | ||
XM_022407429.1 | XP_022263137.1 | 2 - 3 | 315 | 64 | ||
GenBank mRNA | DN344388.1 | - | 2 - 3 | 284 | 64 | |
JQ173134.1 | - | 1 - 2 | 131 | 64 | ||
JQ173135.1 | - | 1 - 2 | 131 | 64 | ||
JQ173136.1 | - | 1 - 2 | 131 | 64 | ||
JQ173137.1 | - | 1 - 2 | 131 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |