Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005633430.3 | XP_005633487.1 | 3 - 4 | 435 | 100 | |
XM_005633431.3 | XP_005633488.1 | 3 - 4 | 374 | 100 | ||
XM_022407587.1 | XP_022263295.1 | 3 - 4 | 333 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |