Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127234.1 | NP_001120706.1 | 1 - 2 | 358 | 90 | |
GenBank mRNA | DN430722.1 | - | 358 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |