Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_533926.6 | XP_533926.1 | 3 - 4 | 357 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |