Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107800.2 | XP_013963275.1 | 356 | 113 | ||
XM_014107801.2 | XP_013963276.1 | 355 | 113 | |||
XM_014107802.2 | XP_013963277.1 | 396 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |