Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534884.5 | XP_534884.1 | 3 - 4 | 488 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |