Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMBIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252399.1 | NP_001239328.1 | 4 - 5 | 360 | 97 | |
XM_005636692.3 | XP_005636749.1 | 4 - 5 | 466 | 97 | ||
XM_022410856.1 | XP_022266564.1 | 4 - 5 | 415 | 97 | ||
GenBank mRNA | DN880529.1 | - | 4 - 5 | 360 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |