Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534901.6 | XP_534901.2 | 2 - 3 | 408 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |