Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535424.6 | XP_535424.1 | 3 - 4 | 680 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |