Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003433752.4 | XP_003433800.1 | 5 - 6 | 1069 | 110 | |
XM_005617874.3 | XP_005617931.1 | 3 - 4 | 537 | 110 | ||
XM_005617875.3 | XP_005617932.1 | 4 - 5 | 527 | 110 | ||
XM_014110941.2 | XP_013966416.1 | 4 - 5 | 813 | 110 | ||
XM_014110945.2 | XP_013966420.1 | 5 - 6 | 820 | 110 | ||
XM_014110954.2 | XP_013966429.1 | 5 - 6 | 784 | 110 | ||
XM_014110967.2 | XP_013966442.1 | 4 - 5 | 563 | 110 | ||
XM_022413847.1 | XP_022269555.1 | 4 - 5 | 575 | 110 | ||
XM_022413880.1 | XP_022269588.1 | 5 - 6 | 576 | 110 | ||
XM_022413884.1 | XP_022269592.1 | 4 - 5 | 465 | 110 | ||
XM_022413888.1 | XP_022269596.1 | 6 - 7 | 880 | 110 | ||
XM_535382.6 | XP_535382.3 | 4 - 5 | 585 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |