Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RORA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535503.7 | XP_535503.3 | 5 - 6 | 908 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |