Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286232.1 | NP_001273161.1 | 3 - 4 | 192 | 118 | |
GenBank mRNA | DN392943.1 | - | 3 - 4 | 192 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |