Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111121.2 | XP_013966596.1 | 3 - 4 | 461 | 142 | |
XM_014111122.2 | XP_013966597.1 | 3 - 4 | 322 | 142 | ||
XM_022415054.1 | XP_022270762.1 | 3 - 4 | 322 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |