Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASNSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001357114.1 | NP_001344043.1 | 4 - 5 | 1758 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |