Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640502.3 | XP_005640559.1 | 3 - 4 | 2751 | 96 | |
XM_022415139.1 | XP_022270847.1 | 3 - 4 | 2938 | 96 | ||
XM_536020.6 | XP_536020.4 | 3 - 4 | 2932 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |