Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014108709.2 | XP_013964184.1 | 2 - 3 | 829 | 89 | |
XM_543766.6 | XP_543766.2 | 3 - 4 | 647 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |