Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5IF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_860617.5 | XP_865710.1 | 1 - 2 | 157 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |