Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RELA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631472.3 | XP_005631529.1 | 3 - 4 | 286 | 60 | |
XM_005631473.2 | XP_005631530.1 | 3 - 4 | 333 | 60 | ||
XM_005631474.3 | XP_005631531.1 | 3 - 4 | 311 | 60 | ||
XM_014121307.2 | XP_013976782.1 | 3 - 4 | 285 | 60 | ||
XM_022405652.1 | XP_022261360.1 | 3 - 4 | 316 | 60 | ||
XM_540850.6 | XP_540850.2 | 3 - 4 | 308 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |