Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616548.3 | XP_005616605.1 | 2 - 3 | 1225 | 65 | |
XM_005616549.3 | XP_005616606.1 | 2 - 3 | 1294 | 65 | ||
XM_005616550.3 | XP_005616607.1 | 2 - 3 | 1196 | 65 | ||
XM_541609.6 | XP_541609.1 | 2 - 3 | 1176 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |