Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_537494.6 | XP_537494.2 | 4 - 5 | 1093 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons |