Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145120.1 | NP_001138592.1 | 8 - 9 | 925 | 95 | |
XM_005623198.3 | XP_005623255.1 | 7 - 8 | 964 | 95 | ||
XM_022421752.1 | XP_022277460.1 | 7 - 8 | 962 | 95 | ||
XM_022421753.1 | XP_022277461.1 | 7 - 8 | 959 | 95 | ||
XM_022421754.1 | XP_022277462.1 | 7 - 8 | 959 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |