Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629370.3 | XP_005629427.1 | 3 - 4 | 747 | 79 | |
XM_022403533.1 | XP_022259241.1 | 3 - 4 | 785 | 79 | ||
XM_022403534.1 | XP_022259242.1 | 3 - 4 | 785 | 79 | ||
XM_022403535.1 | XP_022259243.1 | 3 - 4 | 785 | 79 | ||
XM_022403536.1 | XP_022259244.1 | 3 - 4 | 785 | 79 | ||
XM_532699.5 | XP_532699.1 | 3 - 4 | 785 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |