Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639455.4 | XP_003639503.1 | 2 - 3 | 247 | 116 | |
XM_022426948.1 | XP_022282656.1 | 3 - 4 | 741 | 116 | ||
XM_022426949.1 | XP_022282657.1 | 4 - 5 | 541 | 116 | ||
XM_022426950.1 | XP_022282658.1 | 4 - 5 | 519 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |