Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_531842.6 | XP_531842.1 | 2 - 3 | 284 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |